Learn More
Pyramex™ Rendezvous™ Safety Glasses
Shop All Pyramex Safety Products Products
Click to view available options
Frame Color:
Black
Blue
Crystal Gray
Lens Coating:
Scratch-resistant
Lens Tint:
Amber
Clear
Gray
Indoor/Outdoor Mirror
Infinity Blue
Silver Mirror
Description
Ratcheting lens pitch adjustment aligns lens angle to properly position for work application.
- Flexible nylon temples with four size settings to accommodate a broad range of facial sizes
- Adjustable nose pad conforms to a broad range of bridge sizes
- Lightweight nylon frame for comfortable, all day wear
- Replaceable, 9.5 base-curved polycarbonate lens withstands high-impact
- Provide 99% UV protection, is completely distortion free
Compliance
Meet ANSI Z87.1 requirements.
Specifications
Specifications
Lens Coating | Scratch-resistant |
Certifications/Compliance | Meet ANSI Z87.1 requirements. |
Lens Tint | Clear |
Frame Color | Black |
Temple Style | Flexible |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |