Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyramex™ Lanyard
Shop All Pyramex Safety Products Products
Click to view available options
Product Type:
Breakaway Retainer
Clip-on Retaining Cord
Cotton Retaining Cord
Retaining Cord with Plastic Clips
Description
Fits a variety of temples.
Specifications
Specifications
Color | Black |
Product Type | Retaining Cord with Plastic Clips |
For Use With (Application) | Retaining spectacles |
Includes | Plastic clips |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction