Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Spectacle Storage Cases

Catalog No. 19151628 Shop All Pyramex Safety Products Products
Click to view available options
Product Type:
Eyewear Case
Color:
Black
Blue
For Use With (Application):
For storing glasses
Ideal for lens cleaning
Description:
Drawstring Storage Case
Zippered Storage Case
This item is not returnable. View return policy
This item is not returnable. View return policy

Microfiber cases for glasses ideal for lens cleaning

  • Available in black
  • Available with zipper or drawstring

Specifications

Description Drawstring Storage Case
Color Black
Product Type Eyewear Case
For Use With (Equipment) For glasses
For Use With (Application) For storing glasses
Includes Belt loop
Material Microfiber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.