Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyranose Oxidase Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990315
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0315 100 μg
50-199-0316 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0315 Supplier Rockland Immunochemicals Supplier No. 2001012350100
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-PYRANOSE OXIDASE is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum as well as purified and partially purified Pyranose Oxidase [E.coli]. Cross reactivity against Pyranose Oxidase from other sources is unknown.

Pyranose Oxidase catalyzes the oxidation of various aldopyranoses and disaccharides on carbon-2 to the corresponding 2-keto sugars concomitant with the reduction of O2 to H2O2. It plays an important role in lignin degradation of wood rot fungi by supplying the essential cosubstrate H2O2 for the ligninolytic peroxidases, lignin peroxidase, and manganese-dependent peroxidase. The preferred substrate is D-glucose which is converted to 2-dehydro-D-glucose. Acts also on D-xylose, together with D-glucose the major sugars derived from wood, on L-sorbose, D-galactose and 1,5-anhydroglucitol, a diagnostic marker of diabetes mellitus.

Specifications

Antigen Pyranose Oxidase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Goat
Immunogen Pyranose Oxidase [E.coli]
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.