Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pyrantel pamoate, 98%

Catalog No. AC461890250
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 22204-24-6 | C11H14N2S·C23H16O6 | 594.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22204-24-6
Molecular Formula C11H14N2S·C23H16O6
Molecular Weight (g/mol) 594.68
InChI Key AQXXZDYPVDOQEE-MXDQRGINSA-N
PubChem CID 5281033
ChEBI CHEBI:8655

Specifications

Quantity 25 g
Solubility Information Solubility in water: insoluble
Formula Weight 594.68
Percent Purity 98%
Chemical Name or Material Pyrantel pamoate
EINECSNumber 244-837-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.