Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazinamide, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Plastic bottle
98-96-4
C5H5N3O
123.11
MFCD00006132
IPEHBUMCGVEMRF-UHFFFAOYSA-N
pyrazinamide,pyrazinecarboxamide,zinamide,pyrazinoic acid amide,2-pyrazinecarboxamide,aldinamide,aldinamid,pirazinamid,pyrazineamide,pyrafat
1046
CHEBI:45285
pyrazine-2-carboxamide
C1=CN=C(C=N1)C(=O)N
98-96-4
C5H5N3O
123.11
MFCD00006132
IPEHBUMCGVEMRF-UHFFFAOYSA-N
pyrazinamide,pyrazinecarboxamide,zinamide,pyrazinoic acid amide,2-pyrazinecarboxamide,aldinamide,aldinamid,pirazinamid,pyrazineamide,pyrafat
1046
CHEBI:45285
pyrazine-2-carboxamide
C1=CN=C(C=N1)C(=O)N
Infrared Spectrum Authentic
Color White
Quantity 10g
Melting Point 188°C to 191°C
Packaging Plastic bottle
Assay Percent Range 98.5% min. (HPLC)
Merck Index 15, 8067
Solubility Information Solubility in water: 15mg/mL. Other solubilities: 13.8mg/mL methanol- 5.7mg/mL abs. ethanol,3.8mg/mL isopropanol- 1.0mg/mL ether,0.01mg/mL isooctane- 7.4mg/mL chloroform
Formula Weight 123.11
Physical Form Crystalline Powder or Needles
Percent Purity 99%
Chemical Name or Material Pyrazinamide, 99%
Show More Show Less

EINECSNumber : 202-717-6

RTECSNumber : UQ2275000

TSCA : TSCA