Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazine-2,5-dicarboxylic acid, 95%

Catalog No. AAH6479503
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 122-05-4 | C6H4N2O4 | 168.11 g/mol

It is an pyrazine used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pyrazine used for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-05-4
Molecular Formula C6H4N2O4
Molecular Weight (g/mol) 168.11
MDL Number MFCD00216919
InChI Key GMIOYJQLNFNGPR-UHFFFAOYSA-N
Synonym 2,5-pyrazinedicarboxylic acid, 2,5-pyrazinedicarboxylicacid, pubchem8868, 2,5-pyrazine-diacid, ksc177q1r, pyrazinedicarboxylic-2,5 acid
PubChem CID 255280
IUPAC Name pyrazine-2,5-dicarboxylic acid
SMILES OC(=O)C1=CN=C(C=N1)C(O)=O

Specifications

Melting Point 255°C to 260°C (literature)
Quantity 1 g
Beilstein 139985
Solubility Information Slightly soluble in water.
Formula Weight 168.11
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Pyrazine-2,5-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.