Learn More
Pyrazine, 98%
Description
Pyrazine is used as a flavoring agent. It serves as a component of some herbs in traditional Chinese medicine. It is employed as pharmaceutical intermediates, perfume intermediates and involved in the preparation of other chemicals. In addition to this, it is used in the production of pyrazine 1-oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 50°C to 56°C |
| Color | White |
| Density | 1.031 g/mL |
| Boiling Point | 113°C to 116°C |
| Flash Point | 55°C (131°F) |
| Odor | Strong |
| Refractive Index | 1.5235 |
| Quantity | 50 g |
| UN Number | UN1325 |
| Beilstein | 103905 |
| Show More |
Safety and Handling
| Hazard Category | H228-H315-H319-H335 |
| Hazard Statement | GHS H Statement H228-H315-H319-H335 Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 206-027-6 |
| RTECSNumber | UQ2015000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.