Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazine, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
290-37-9
C4H4N2
80.09
KYQCOXFCLRTKLS-UHFFFAOYSA-N
1,4-diazine,p-diazine,paradiazine,piazine,1,4-diazabenzene,pyrazin,ccris 1331,1,4-diazin,mixture,mu-pyrazine;
9261
CHEBI:30953
pyrazine
C1=CN=CC=N1
290-37-9
C4H4N2
80.09
KYQCOXFCLRTKLS-UHFFFAOYSA-N
1,4-diazine,p-diazine,paradiazine,piazine,1,4-diazabenzene,pyrazin,ccris 1331,1,4-diazin,mixture,mu-pyrazine;
9261
CHEBI:30953
pyrazine
C1=CN=CC=N1
Flash Point 55°C
Boiling Point 115.0°C to 116.0°C
Color White
Packaging Glass bottle
Melting Point 52.0°C to 56.0°C
Infrared Spectrum Authentic
Quantity 100g
Assay Percent Range 99+%
Beilstein 23,91
Merck Index 15,8068
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol and ether
Formula Weight 80.09
Physical Form Crystals
Percent Purity ≥99%
Chemical Name or Material Pyrazine
Show More Show Less