Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazole-3-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. P22611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P22611G 1 g
P2261200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P22611G Supplier TCI America Supplier No. P22611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3920-50-1
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.09
MDL Number MFCD00129925
InChI Key ICFGFAUMBISMLR-UHFFFAOYSA-N
Synonym 3-Formylpyrazole
PubChem CID 12218383
IUPAC Name 1H-pyrazole-5-carbaldehyde
SMILES O=CC1=CC=NN1

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 1 g
Formula Weight 96.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pyrazole-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.