Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrazole, 98%, pure

Catalog No. p-188907
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 288-13-1 | C3H4N2 | 68.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 288-13-1
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.08
MDL Number MFCD00005234
InChI Key WTKZEGDFNFYCGP-UHFFFAOYSA-N
Synonym pyrazole, 1,2-diazole, 1h-pyrazol, pyrazol, unii-3qd5kjz7zj, 3qd5kjz7zj, diazole, hpz, 1-h-pyrazole, pyrazol#1
PubChem CID 1048
ChEBI CHEBI:17241
IUPAC Name 1H-pyrazole
SMILES C1=CNN=C1

Specifications

Melting Point 66.0°C to 70.0°C
Color White to Yellow
Boiling Point 186.0°C to 188.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 23, 39
Fieser 01,958; 12,419
Merck Index 15, 8071
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 68.08
Percent Purity 98%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pyrazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-017-1
RTECSNumber UQ4900000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.