Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyrethroid hydrolase Ces2e Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306082AF594
Description
Pyrethroid hydrolase Ces2e Polyclonal specifically detects Pyrethroid hydrolase Ces2e in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Pyrethroid hydrolase Ces2e | |
Polyclonal | |
Western Blot, ELISA | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Mouse | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Produced in rabbits immunized with purified, recombinant Mouse Pyrethroid hydrolase Ces2e (NP_766347.1; Met1-His556). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction