Learn More
Description
Includes
Unground glass tube (mortar) and two pestles
Supplied with two pestles to ensure dissociation of cells into fine particles with minimal damage to cell nuclei. Use the large clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided.
Specifications
Specifications
| Sterility | Non-sterile |
| Capacity (English) | 0.5 oz. |
| Capacity (Metric) | 15 mL |
| Material | Borosilicate Glass |
| Product Type | Dounce Tissue Grinder |
| For Use With (Application) | Ideal for Enzyme studies where heat build-up must be avoided, use large clearance pestle for initial reduction of soft tissue, use small clearance pestle to complete homogenization |
| Includes | Mortar tube, large pestle and small pestle |
| Length (English) Tube | 5.7 in. |
| Length (Metric) Tube | 145 mm |
| Tube Diameter (English) Outside | 0.78 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.