PYREX™ Glass Chromatographic Reagent Atomizer Available on GSA/VA Contract for Federal Government customers only.

Used for spraying reagents onto thin layer chromatography plates

Manufacturer: Corning, Inc.Supplier Diversity Partner 2153250

 View more versions of this product

Catalog No. 05-723-26B

  Check Availability
Add to Cart

    Specifications

    Product Type Chromatographic reagent atomizer
    Capacity (English) 8.45 oz.
    Capacity 8.45 oz., 250mL
    For Use With (Application) For spraying reagents in thin layer chromatography
    Height (English) 8.18 in.
    Capacity (Metric) 250mL
    Height (Metric) 20.8cm
    Includes Full Length 24/40 joint
    Material Pyrex Glass
    Quantity 1 Case
    Standard Taper Joint 24/40

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit