Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ PYREX™ Large-Capacity Bottles with Tooled Neck
Shop All Corning Products
Click to view available options
Capacity (Metric):
13,200 mL
19,000 mL
45,500 mL
9000 mL
9500 mL
Graduations:
No
Yes
Description
For distilled water and solutions
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Order Info
Bottles take No. 12 size stoppers (not included).
Specifications
Specifications
| Capacity | 448 oz., 13200 mL |
| Capacity (Metric) | 13,200 mL |
| Capacity (English) | 448 oz. |
| Color | Clear |
| Diameter (English) | 9.37 in. |
| Material | Borosilicate Glass |
| No. per Case | 4/Cs. |
| No. per Pack | 4/Pk. |
| Sterility | Non-sterile |
| Graduations | Yes |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction