Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Large-Capacity Bottles with Tooled Neck
BIOPHARMAGSA_VA

Catalog No. 02887A Shop All Corning Products
Click to view available options
Capacity (Metric):
13,200 mL
19,000 mL
45,500 mL
9000 mL
9500 mL
Graduations:
No
Yes

For distilled water and solutions

  • Individual alphanumerical codes for well identification, flat bottoms
  • Treated for optimal cell attachment (except where noted)
  • Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
  • Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
  • Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
  • Sterile
  • Nonpyrogenic

Order Info

Bottles take No. 12 size stoppers (not included).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 320 oz., 9500 mL
Capacity (Metric) 9500 mL
Capacity (English) 320 oz.
Diameter (English) 7.3 in.
Material Borosilicate Glass
No. per Case 4/Cs.
No. per Pack 4/Pk.
Sterility Non-sterile
Graduations No
Shape Cylindrical
Height (Metric) 470 mm
Height (English) 18.5 in.
Diameter (Metric) 187 mm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.