Learn More
Corning™ PYREX™ Large-Capacity Bottles with Tooled Neck
Shop All Corning Products

Description
For distilled water and solutions
- Individual alphanumerical codes for well identification, flat bottoms
- Treated for optimal cell attachment (except where noted)
- Corning™ CellBIND™ surface is a novel cell culture treatment that increases surface wettability for more even and consistent cell attachment.
- Ultra-Low Attachment surface plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption, and cellular activation.
- Corning Osteo Assay surface is an inorganic crystalline coating, creating a surface that mimics in vivo bone-like, for in vitro bone cell culture and assays.
- Sterile
- Nonpyrogenic
Order Info
Bottles take No. 12 size stoppers (not included).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Capacity | 294.4 oz., 9000 mL |
| Capacity (Metric) | 9000 mL |
| Capacity (English) | 294.4 oz. |
| Diameter (English) | 7.3 in. |
| Material | Borosilicate Glass |
| No. per Case | 4/Cs. |
| No. per Pack | 4/Pk. |
| Sterility | Non-sterile |
| Graduations | Yes |
| Shape | Cylindrical |
| Show More |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.