Learn More
PYREX™ Liebig Condensers
For vacuum distillation
Manufacturer:  Corning 2400400
 View more versions of this product
Catalog No. 07-721C
Add to cart
Description
Description
For vacuum distillation
  • standard taper outer joint at top
  • standard taper inner joint with drip tip at bottom
  • Both standard taper joints are same size
  • Hose connections are 10mm O.D.
Specifications
Specifications
For use in vacuum distillations
550mm
400mm
24/40
21.65 in.
15.74 in.
Liebig Condenser