Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Polyethylene Snap Caps for Volumetric Flasks
GSA_VAGREENER_CHOICE

Catalog No. p-3318945
Click to view available options
Closure Size:
10
100
1000
200
2000
25
250
50 mm; Stopper No. 50
500
This item is not returnable. View return policy
This item is not returnable. View return policy

Economical closure for some volumetric flasks

These blue caps provide an economical closure for certain volumetric flasks. Refer to volumetric flask listings to determine sizes and whether the flask is designed to take such a cap.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Closure Color Blue
Closure Material HDPE
Color Blue
For Use With (Equipment) For volumetric flasks
No. per Case 6/Cs.
Product Type Cap, Snap
Disposable Reusable
Closure Type Snap Cap
Closure Size 100
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.