Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Corning™ PYREX™ Reagent Bottles with Hollow standard taper Stoppers
Shop All Corning Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1 L
2000 mL
Stopper Number:
19
24
29
Description
With neck ground and furnished with standard taper stopper.
For stopper only, see Cat. No. 7575.
Order Info
Replacement stoppers available (No. 7575 series).

Specifications
Specifications
Capacity | 8.45 oz., 250 mL |
Capacity (Metric) | 250 mL |
Capacity (English) | 8.45 oz. |
Diameter (English) | 2.75 in. |
Diameter (English) Outer Neck | 1.22 in. |
Diameter (Metric) Outer Neck | 31 mm |
Material | Borosilicate Glass |
No. per Case | 36/Cs. |
No. per Pack | 1/Pk. |
Sterility | Non-sterile |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction