Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Reagent Bottles with PYREX Hollow standard taper Stoppers
SCI_ED

Catalog No. S31247 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
500 mL
Stopper Number:
24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Stopper Number
S31247 500 mL 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S31247 Supplier Corning™ Supplier No. 1500500/EMD

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Plain, narrow mouth bottles with ground necks and hollow glass stoppers.

  • Well-suited for storing reagents, sera, vaccines, colloidal solutions and similar liquids

TRUSTED_SUSTAINABILITY

Specifications

Capacity 16.9 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Diameter (English) 3.38 in.
Diameter (English) Outer Neck 1.39 in.
Diameter (Metric) Outer Neck 35.5 mm
Material Glass
Sterility Non-sterile
Stopper Number 24
Mouth Narrow
Shape Cylindrical
Height (Metric) 198 mm
Height (English) 7.79 in.
Diameter (Metric) 86 mm
Closure Type Hollow Glass Stopper
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.