Learn More
Description
Ideal for plant tissue culture and general culture work. Available in packs of 36 except S67135 which is available in packs of 24.
• Walls and bottoms are uniform thickness
• Rimless top for greater ease in plugging and rack storage
• Flat bottom facilitates use with agar slants
Two-position polypropylene closures available separately. Available in cases of 100.
• Autoclavable
• Position closure OPEN for gas exchange or CLOSED for humidified environment
• Internal drip ring minimizes contamination
Specifications
Specifications
| Capacity (Metric) | 39 mL |
| Length (Metric) | 100 mm |
| Material | Borosilicate Glass |
| Disposable | Reusable |
| For Use With | For plant tissue culture and general culture work |
| Label Type | None |
| Rimmed | No |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| Graduated | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
