Learn More
Corning™ PYREX™ Hollow Glass Standard Taper Stoppers
GREENER_CHOICE
Catalog No. p-78313 Shop All Corning Products
Change view
Click to view available options
Size:
8
9
13
16
19
22
27
32
38
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
14-640-1F 27
14-640-1B 13
14-640-1E 22
14-640-1A 9
14-640-1AA 8
14-640-1G 32
14-640-1H 38
14-640-1C 16
14-640-1D 19
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14-640-1F Supplier Corning™ Supplier No. 765027
Only null left
Add to Cart
Add to Cart
Variety of sizes available
The hollow stopper is light in weight, yet very strong to reduce chipping and breakage, thus minimizing replacement costs. A barrel-shaped head means it is easy to clean and even easier to rotate in or out. Grooves in the sides help prevent slippage. The flat top allows the stopper to stand on its head, which frees the user’s hand and minimizes potential contamination. This stopper is interchangeable with other common flask stoppers and fits all standard labware of comparable standard taper size.
TRUSTED_SUSTAINABILITY
Specifications
Size 27
For Use With (Equipment) Flask and separatory funnel
Height (Metric) 5.4 cm
Length (Metric) Ground Zone 2.15 cm
Material Borosilicate Glass
Product Type Barrelhead Stopper