Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ PYREX™ Water Sampling/Reagent Bottle

Catalog No. 03374 Shop All Corning Products
Click to view available options
Capacity (Metric):
125 mL

Cylindrical, wide-mouth reagent bottle

 

  • With hollow glass standard taper stopper
  • Height without stopper: 108 mm
  • Height with stopper: 142 mm
  • Heavy wall
  • Reusable
  • Nonsterile

 

Order Info

Replacement stoppers available (No. 7575-29)

Compliance

Meets APHA requirements.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Diameter (English) 2.2 in.
Diameter (English) Outer Neck 1.5 in.
Diameter (Metric) Outer Neck 38.25 mm
Includes Interchangeable stopper ground
Material Borosilicate Glass
No. per Case 24/Cs.
No. per Pack 12/Pk.
Sterility Non-sterile
Stopper Number 29
Certifications/Compliance APHA
Mouth Wide
Shape Cylindrical
Height (Metric) 142 mm
Height (English) 5.5 in.
Closure Material Polypropylene
Diameter (Metric) 57 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.