Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-2-carboxylic Acid Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P042225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P042225G 25 g
P04225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P042225G Supplier TCI America Supplier No. P042225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 636-80-6
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.57
MDL Number MFCD00050666
InChI Key HFAFXVOPGDBAOK-UHFFFAOYSA-N
Synonym Picolinic Acid Hydrochloride
PubChem CID 71559
IUPAC Name hydrogen pyridine-2-carboxylic acid chloride
SMILES [H+].[Cl-].OC(=O)C1=CC=CC=N1

Specifications

Color White
Quantity 25 g
Formula Weight 159.57
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2-carboxylic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.