Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
1L
2.5L
Packaging:
AcroSeal™ Glass Bottle
Acroseal Glass Bottle
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene,azine,pyridin,piridina,pirydyna,pyridin german,pirydyna polish,piridina italian,rcra waste number u196,caswell no. 717
1049
CHEBI:16227
pyridine
C1=CC=NC=C1
110-86-1
C5H5N
79.102
MFCD00011732
JUJWROOIHBZHMG-UHFFFAOYSA-N
azabenzene,azine,pyridin,piridina,pirydyna,pyridin german,pirydyna polish,piridina italian,rcra waste number u196,caswell no. 717
1049
CHEBI:16227
pyridine
C1=CC=NC=C1
Additional Information Vapor Pressure: 20mbar at 20°C
Assay Percent Range 99.5%
Beilstein 20, 181
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Merck Index 15, 8081
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Formula Weight 79.1
Physical Form Liquid
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Density 0.9780g/mL
Infrared Spectrum Authentic
Packaging Acroseal Glass Bottle
Refractive Index 1.5085 to 1.5105
Specific Gravity 0.978
Water 0.005% Max.
Flash Point 17°C
Viscosity 0.95 mPa.s (20°C)
Boiling Point 115.0°C to 116.0°C
Color Colorless
Melting Point -42.0°C
Quantity 100mL
Chemical Name or Material Pyridine, Over Molecular Sieves
Show More Show Less

GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger