Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, 99+%

Catalog No. p-7081840
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Catalog No. Quantity
AAA12005AP 500 mL
AAA120050F 2500 mL
AAA120050D 10,000 mL
3 options

Catalog No. AAA12005AP

Supplier: Thermo Scientific Chemicals A12005AP

Only null left
Add to Cart

CAS: 110-86-1 | C5H5N | 79.102 g/mol

As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane and pyridine-sulfur trioxide are widely used owing to their superior characteristics in their reactivity and solubility characteristics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As polar, basic, low-reactive solvent, precursor, and reagent, pyridine finds wide range of applications in pharmaceuticals, agrochemicals and in the manufacture of dyes and rubber. Further it is used in the textile industry to improve network capacity of cotton. Pyridine finds applications in specialty chemical reagents like Cornforth reagent (pyridinium dichromate, PDC), pyridinium chlorochromate (PCC), the Collins reagent (complex of chromium(VI) oxide with pyridine) and the Sarret reagent (complex of chromium(VI) oxide with pyridine). Pyridine is a widely used ligand in coordination chemistry. Pyridine-base adducts like pyridine-borane and pyridine-sulfur trioxide are widely used owing to their superior characteristics in their reactivity and solubility characteristics.

Solubility
Miscible with water, alcohol, ether, pet ether, and oils.

Notes
Pyridine is stable under normal temperatures and pressures. It is incompatible with strong oxidizing agents, and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point °C to 42°C
Density 0.978
Boiling Point 115°C
Quantity 500 mL
Flash Point 20°C (68°F)
Physical Form Liquid
Refractive Index 1.51
Assay Percent Range ≥99%
UN Number UN1282
Beilstein 103233
Merck Index 14,7970
Solubility Information Miscible with water,alcohol,ether,pet ether,and oils.
Formula Weight 79.1
Percent Purity ≥99%
Chemical Name or Material Pyridine
Show More Show Less
Hazard Category H225-H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H225-H302-H312-H332
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRIDINE
EINECSNumber 203-809-9
RTECSNumber UR8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.