Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, 99+%, ACS reagent

Catalog No. p-216731
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Metal Drum

CAS: 110-86-1 | C5H5N | 79.102 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Melting Point -42.0°C
Color Colorless
Density 0.9780g/mL
Boiling Point 115.0°C to 116.0°C
Quantity 500 mL
Flash Point 17°C
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.5085 to 1.5105
Residue after Evaporation 0.002% max.
Assay Percent Range 99+%
Specific Gravity 0.978
Vapor Pressure Vapor Pressure: 20mbar at 20°C
UN Number 1282
Viscosity 0.95 mPa.s (20°C)
Beilstein 20, 181
Water 0.1% max. (K.F.)
Fieser 01,958; 02,349; 04,414; 06,497; 11,448; 12,416; 13,293; 15,1; 17,153
Merck Index 15, 8081
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and most other common, organic solvents
Formula Weight 79.1
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material Pyridine
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-809-9
RTECSNumber UR8400000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.