Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyridine, 99+%, for spectroscopy
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy

Specifications
Specifications
Melting Point | -42.0°C |
Absorbance | 0.022 max. at 345nm, 0.097 max. at 340nm, 0.301 max. at 335nm, 1 max. at 310nm |
Density | 0.9780g/mL |
Boiling Point | 115.0°C to 116.0°C |
Quantity | 1 L |
Flash Point | 17°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.5085 to 1.5105 |
Assay Percent Range | 99+% |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS P Statement Harmful in contact with skin. Harmful if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. Highly flammable liquid and vapor. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 203-809-9 |
RTECSNumber | UR8400000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction