Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pyridine N-oxide, 95%

Manufacturer:  Alfa Aesar™ A1041936

Quantity 500g

 View more versions of this product

Catalog No. AAA1041936


  Check Availability
Add to cart

Specifications

Description

Solubility
Soluble in water.

Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
Description & Specifications

Chemical Identifiers

694-59-7
95.1
105257
Pyridine N-oxide
C5H5NO
MFCD00006194
14,7972

Specifications

143°C (289°F)
61° to 66°C
694-59-7
95.1
105257
Pyridine N-oxide
270°C
500g
C5H5NO
MFCD00006194
14,7972
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : UT6410000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit