Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-N-oxide, 95%

Catalog No. p-189638
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 694-59-7 | C5H5NO | 95.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-59-7
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.1
MDL Number MFCD00006194
InChI Key ILVXOBCQQYKLDS-UHFFFAOYSA-N
Synonym pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide
PubChem CID 12753
ChEBI CHEBI:29136
IUPAC Name 1-oxidopyridin-1-ium
SMILES C1=CC=[N+](C=C1)[O-]

Specifications

Melting Point 60.0°C to 65.0°C
Color Brown to White
Boiling Point 270.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Beilstein 20, II, 131
Fieser 01,966; 02,353; 05,567; 09,396
Merck Index 15, 8084
Solubility Information Solubility in water: soluble.
Formula Weight 95.1
Percent Purity 95%
Physical Form Crystalline Solid, Crystals, or Chunks
Chemical Name or Material Pyridine-N-oxide, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-774-6
RTECSNumber UT6410000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.