Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridinium bromide perbromide, 90+%, technical

Catalog No. ac158361000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

CAS: 39416-48-3 | C5H6Br3N | 319.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39416-48-3
Molecular Formula C5H6Br3N
Molecular Weight (g/mol) 319.83
MDL Number MFCD00013223
Synonym Pyridine hydrobromide, compound with bromine

Specifications

Melting Point 127°C to 133°C
Color Brown to Orange-Red
Infrared Spectrum Authentic
Assay Percent Range 90% min. (Iodometry)
Packaging Glass bottle
Quantity 100 g
Beilstein 20, V,5, 181
Fieser 01,967; 05,568; 06,499; 09,399; 10,333; 14,268; 15,276
Merck Index 15, 8085
Solubility Information Solubility in water: decomposes. Other solubilities: sparingly soluble in acetic acid
Formula Weight 319.83
Percent Purity 90+%
Grade Technical
Physical Form Powder, Crystals or Chunks
Chemical Name or Material Pyridinium bromide perbromide, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 254-446-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.