Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridinium p-toluenesulfonate, 98%

Catalog No. AC213740050
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 24057-28-1 | C12H13NO3S | 251.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24057-28-1
Molecular Formula C12H13NO3S
Molecular Weight (g/mol) 251.3
MDL Number MFCD00013108
Synonym pyridinium p-toluenesulfonate, ppts, pyridin-1-ium 4-methylbenzenesulfonate, pyridinium 4-toluenesulfonate, pyridinium tosylate, pyridinium 4-methylbenzenesulfonate, pyridinium p-toluene sulfonate, pyridinium p-toluene sulphonate, pyridinium para-toluenesulfonate, 4-toluenesulfonic acid pyridine salt

Specifications

Melting Point 116.0°C to 120.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 20, V,5, 160
Fieser 08,427; 09,400; 12,420; 15,276; 16,287
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ethanol, acetone, diethyl ether
Formula Weight 251.3
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pyridinium p-toluenesulfonate
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Warning
missing translation for 'healthHazard2' GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 246-002-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.