Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Pyridoin, 97%, Thermo Scientific™

Catalog No. AC345620250
Click to view available options
Quantity:
25 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1141-06-6
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
MDL Number MFCD00010691
InChI Key ZKBDAJDDDOIASC-UHFFFAOYSA-N
Synonym alpha-pyridoin, .alpha.-pyridoin, ethanone, 2-hydroxy-1,2-di-2-pyridinyl, ketone, hydroxy-2-pyridylmethyl 2-pyridyl, 2-hydroxy-1,2-bis pyridin-2-yl ethanone, a-pyridoin, 2-hydroxy-1,2-di 2-pyridyl ethan-1-one, 2-hydroxy-1,2-di-2-pyridylethanone, ethanone,2-di-2-pyridinyl
PubChem CID 95624
IUPAC Name 2-hydroxy-1,2-dipyridin-2-ylethanone
SMILES C1=CC=NC(=C1)C(C(=O)C2=CC=CC=N2)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 151°C to 158°C
Color Brown to Brown-Red
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 25 g
Beilstein 25, I, 475
Formula Weight 214.22
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material α-Pyridoin, 97%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.