Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Pyridostatin pentahydrochloride

Catalog No. 476310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-631-0 10 mg
47-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-631-0 Supplier Tocris Bioscience™ Supplier No. 4763/10
This item is not returnable. View return policy

Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest

Binds and stabilizes G-quadruplexes, inducing DNA damage and cell cycle arrest (Kd = 490 nM); targets the proto-oncogene Src, reducing Src protein abundance and Src-dependent motility in human breast cancer cells. Also targets telomeric G-quadruplexes, inducing telomerase dysfunction. Activates the DNA-dependent protein kinase catalytic sunbunit (DNA-PKcs).

Specifications

CAS 1781882-65-2
Quantity 10 mg
Molecular Formula 4-(2-Aminoethoxy)N2,N6-bis[(4-(2-aminoethoxy)-2-quinolinyl]-2,6-pyridinecarboxamide pentahydrochloride
Product Type Pyridostatin pentahydrochloride
Purity 95%
Molecular Weight (g/mol) M.W. (Observed): 778.94
Inhibitors Pyridostatin pentahydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.