Learn More
Pyridoxal-5-phosphate hydrate, 98%
Description
Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Active form of Vitamin B-6Pyridoxal-5’-phosphate monohydrate is used to modify the lysyl and valyl residues in proteins. It acts as a coenzyme in transamination reactions and as an inhibitor for purinergic receptors and intracellular. It is involved in decarboxylation and deamination reactions viz. the conversion of dopa into dopamine of glutamate into GABA and histidine to histamine.
Solubility
Soluble in alkaline solutions. Sparingly soluble in water.
Notes
Light sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.

Specifications
Specifications
Melting Point | 142°C |
Quantity | 5 g |
Sensitivity | Light sensitive |
Merck Index | 14,7979 |
Solubility Information | Soluble in alkaline solutions. Sparingly soluble in water. |
Formula Weight | 265.17 (247.15 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | Pyridoxal-5-phosphate hydrate |
Safety and Handling
EINECSNumber | 200-208-3 |
RTECSNumber | UV1207000 |
TSCA | Yes |
Recommended Storage | Store at -20°C; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.