Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pyridoxal Kinase/PDXK Antibody (OTI5H5), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273754PE
Description
Pyridoxal Kinase/PDXK Monoclonal specifically detects Pyridoxal Kinase/PDXK in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
Pyridoxal Kinase/PDXK | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
PDXK | |
Full length recombinant protein of human Pyridoxal Kinase/PDXK (NP_03672) produced in HEK293 cell. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry | |
OTI5H5 | |
Flow Cytometry | |
C21orf124, C21orf97, chromosome 21 open reading frame 124, DKFZp566A071, EC 2.7.1.35, FLJ21324, FLJ37311, MGC15873, MGC31754, MGC52346, PKHchromosome 21 open reading frame 97, PNKhuman pyridoxal kinase, EC 2.7.1.3510FLJ31940, PRED79, pyridoxal (pyridoxine, vitamin B6) kinase, pyridoxal kinase, pyridoxamine kinase, Pyridoxine kinase, vitamin B6 kinase | |
Mouse | |
Affinity Purified | |
metabolism | |
8566.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction