Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605995
Click to view available options
Quantity:
100 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

PY103, 58-56-0, C8H11NO3·HCl

Spectrum™ Chemical Pyridoxine Hydrochloride, USP , a form of vitamin B6, is used to prevent or treat peripheral neuropathy nerve disorder. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.64
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
IUPAC Name hydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
SMILES [H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Poly Pail
Quantity 12 kg
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.