Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridoxine Hydrochloride, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18605996 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-996 25 kg Fiber Drum
18-605-993 100 g Amber Glass Bottle
18-605-994 1 kg Amber Glass Bottle
18-605-995 12 kg Poly Pail
18-605-997 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-996 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PY10325KGBL

PY103, 58-56-0, C8H11NO3·HCl

Spectrum™ Chemical Pyridoxine Hydrochloride, USP , a form of vitamin B6, is used to prevent or treat peripheral neuropathy nerve disorder. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 58-56-0
Molecular Formula C8H12ClNO3
Molecular Weight (g/mol) 205.64
InChI Key ZUFQODAHGAHPFQ-UHFFFAOYSA-N
IUPAC Name hydrogen 4,5-bis(hydroxymethyl)-2-methylpyridin-3-ol chloride
SMILES [H+].[Cl-].CC1=NC=C(CO)C(CO)=C1O

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.