Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrimethamine, 100.5%, MP Biomedicals™

Catalog No. ICN19418001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19418001 1 g
ICN19418005 5 g
ICN19418025 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19418001 Supplier MP Biomedicals, Inc Supplier No. 0219418001
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58-14-0
Molecular Formula C12H13ClN4
Molecular Weight (g/mol) 248.71
MDL Number MFCD00057350
InChI Key WKSAUQYGYAYLPV-UHFFFAOYSA-N
Synonym pyrimethamine, chloridine, daraprim, chloridin, diaminopyritamin, 5-4-chlorophenyl-6-ethylpyrimidine-2,4-diamine, ethylpyrimidine, pirimetamina, chloridyn, malocide
PubChem CID 4993
ChEBI CHEBI:8673
IUPAC Name 5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
SMILES CCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1

Specifications

Color White
Assay Percent Range 1.005
Quantity 1 g
Formula Weight 248.7
Physical Form Powder
Chemical Name or Material Pyrimethamine
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.