Learn More
Pyrimidine-5-carboxaldehyde, 97%
Description
Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyrimidine-5-carboxaldehyde is used in the preparation of (S)- and (R)-pyrimidyl alkanol by reacting with diisopropylzinc and (n-butyl)ethyl(n-hexyl)(n-propyl)methane.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 39°C to 43°C |
| Density | 1.23 |
| Boiling Point | 80°C to 82°C (4 mmHg) |
| Quantity | 5 g |
| Sensitivity | Air Sensitive |
| Formula Weight | 108.1 |
| Percent Purity | 97% |
| Chemical Name or Material | Pyrimidine-5-carboxaldehyde |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.