Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pyrimidine, 99%

Click to view available options
Quantity:
1g
5g
25g
289-95-2
C4H4N2
80.09
MFCD00006059
CZPWVGJYEJSRLH-UHFFFAOYSA-N
1,3-diazine,metadiazine,m-diazine,miazine,1,3-diazabenzene,base,1,3-diazin,pyrimidin,unii-k8cxk5q32l,k8cxk5q32l
9260
CHEBI:16898
pyrimidine
C1=CN=CN=C1
289-95-2
C4H4N2
80.09
MFCD00006059
CZPWVGJYEJSRLH-UHFFFAOYSA-N
1,3-diazine,metadiazine,m-diazine,miazine,1,3-diazabenzene,base,1,3-diazin,pyrimidin,unii-k8cxk5q32l,k8cxk5q32l
9260
CHEBI:16898
pyrimidine
C1=CN=CN=C1
Applications
Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
Density 1.016
Refractive Index 1.504
Sensitivity Hygroscopic
Quantity 25g
Boiling Point 123°C to 124°C
Flash Point 31°C (87°F)
Melting Point 20°C to 22°C
UN Number UN1993
Beilstein 103894
Merck Index 14,7987
Solubility Information Fully miscible in water.
Formula Weight 80.09
Percent Purity 99%
Chemical Name or Material Pyrimidine
Show More Show Less

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 206-026-0

RTECSNumber : UV6263000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen