Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrimidine, 98+%

Catalog No. p-7022657
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 289-95-2 | C4H4N2 | 80.09 g/mol

Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis and discovery of antiviral medication, such as in the case of HIV and HSV. Also used in the synthesis of potent inhibitors of 15-lipoxygenase in the reduction of the release of leukotrienes. And in ithe production of RNA and DNA, proteins and starches, the regulation of enzymes and cell signaling.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289-95-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006059
InChI Key CZPWVGJYEJSRLH-UHFFFAOYSA-N
Synonym 1,3-diazine, metadiazine, m-diazine, miazine, 1,3-diazabenzene, base, 1,3-diazin, pyrimidin, unii-k8cxk5q32l, k8cxk5q32l
PubChem CID 9260
ChEBI CHEBI:16898
IUPAC Name pyrimidine
SMILES C1=CN=CN=C1

Specifications

Melting Point 20°C to 22°C
Density 1.016
Boiling Point 123°C to 124°C
Flash Point 31°C (87°F)
Refractive Index 1.504
Quantity 1 g
UN Number UN1993
Beilstein 103894
Sensitivity Hygroscopic
Merck Index 14,7987
Solubility Information Fully miscible in water.
Formula Weight 80.09
Percent Purity 99%
Chemical Name or Material Pyrimidine
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 206-026-0
RTECSNumber UV6263000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.