Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrin, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABF116925UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABF116925UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABF116925UL Supplier MilliporeSigma Supplier No. ABF116925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Pyrin, Cat. No. ABF1169, is a rabbit polyclonal antibody that detects human Pyrin and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Pyrin (UniProt: O15553; also known as Marenostrin) is encoded by the MEFV (also known as MEF, TRIM20) gene (Gene ID: 4210) in human. Pyrin is a homotrimeric protein that is involved in the regulation of innate immunity and the inflammatory response in response to IFNG/IFN-gamma. It organizes autophagic machinery by serving as a platform for the assembly of ULK1, Beclin 1/BECN1, ATG16L1, and ATG8 family members and recognizes specific autophagy targets, thus coordinating target recognition with assembly of the autophagic apparatus and initiation of autophagy. Pyrin acts as an autophagy receptor for the degradation of several inflammasome components, including CASP1, NLRP1 and NLRP3, hence it may prevent excessive IL-1B- and IL-18-mediated inflammation. Pyrin is expressed in peripheral blood leukocytes, particularly in mature granulocytes and to a lesser extent in monocytes, but not in lymphocytes. It is also detected in spleen, lung and muscle, probably as a result of leukocyte infiltration in these tissues. Its expression is also detected in several myeloid leukemic, colon cancer, and prostate cancer cell lines. Three isoforms of Pyrin have been described that are produced by alternative splicing. Isoform 1 associates with microtubules and with the filamentous actin of perinuclear filaments and peripheral lamellar ruffles. In pre-apoptotic cells, it co-localizes with PYCARD/ASC in large specks (inflammasome). Isoform 2 is shown to be localized to the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pyrin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Pyrin in human tonsil, human cerebral cortex, human skeletal muscle, and human pancreatic cancer tissues.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Marenostrin
Gene Symbols MEFV;MEF;TRIM20
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal region of human Pyrin, isoform 1.
Molecular Weight of Antigen ∽82 kDa observed; 86.44 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Pyrin in human cells. It targets an epitope within 15 amino acids from the N-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.