Learn More
Eisco™ Pyrite Mineral Specimen
SCI_ED
Catalog No. S24708 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24708 1 Ea. 2 to 3 cm
S26615 10/Pk. 2 to 3 cm
S24709 12/Pk. 2 to 3 cm
S26655 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24708 Supplier Eisco™ Supplier No. ESNG0024
Add to Cart
Add to Cart
Pyrite Mineral Specimen has a shiny metallic luster and bright brassy color, making it an ideal addition to mineral collections or classroom exercises.
  • Metallic luster and color often lead to mistaking this mineral for gold
  • Leaves behind black streak
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • For replacing other minerals or fossils through pyritization
Specifications
Product Type Mineral
For Use With (Application) Pyritization
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Pyrite