Learn More
Pyrococcus furiosus albA Polyclonal Antibody

Description
Pyrococcus furiosus albA Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | Pyrococcus furiosus albA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | albA |
| Gene Accession No. | Q8TZV1 |
| Gene Alias | DNA/RNA-binding protein Alba |
| Gene Symbols | albA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.