Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Pyrococcus furiosus PF0142 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117693
Description
This antibody has been tested in direct-ELISA.
Putative L-asparaginase performs the following catalytic activity: H2O + L-asparagine = L-aspartate + NH4+.
Specifications
Pyrococcus furiosus PF0142 | |
Polyclonal | |
Unconjugated | |
PF0142 | |
Putative L-asparaginase | |
Rabbit | |
Protein G | |
RUO | |
1467974 | |
-20°C or -80°C if preferred | |
Liquid |
Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8U4E6 | |
PF0142 | |
Recombinant Pyrococcus furiosus Putative L-asparaginase protein (1-175aa). | |
100 μg | |
Primary | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction