Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrogallol, ACS, 99%

Catalog No. p-7059190
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol

Pyrogallol is used in hair dying and suturing materials dying. It is used as a developing agent in black and white developers, as a metal- complexing agent and is involved in oxygen absorption in gas analysis. It exhibits antiseptic properties. Its alkaline solution is used to absorb oxygen from air, thereby it plays a vital role to calculate the amount of oxygen in air.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-66-1
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00002192
InChI Key WQGWDDDVZFFDIG-UHFFFAOYSA-N
Synonym pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
PubChem CID 1057
ChEBI CHEBI:16164
IUPAC Name benzene-1,2,3-triol
SMILES OC1=CC=CC(O)=C1O

Specifications

Melting Point 131°C to 135°C
Color White
Density 1.45 g/mL
Boiling Point 309°C
Odor Odorless
Quantity 10 g
UN Number UN2811
Beilstein 907431
Sensitivity Air and light sensitive
Merck Index 14,8000
Solubility Information Soluble in ethanol,water and ether. Slightly soluble in chloroform.
Formula Weight 126.11
Percent Purity 99%
Grade ACS
Physical Form Powder
Chemical Name or Material Pyrogallol
Show More Show Less
Hazard Category H301-H312+H332-H315-H319-H341
Hazard Statement GHS H Statement
H341-H302-H312-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 201-762-9
RTECSNumber UX2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.