Learn More
Trajan Scientific and Medical Pyrojector SI-1 (N&P) Needle Kit
SDP
Catalog No. SG0316287 Shop All SGE Inc. Products
Change view
Click to view available options
Needle Kit
Catalog No.
SG-0316287 Needle Kit
1 options
Catalog No. SG-0316287 Supplier Trajan Scientific and Medical Supplier No. 0316287
Pyrolysis Gas Chromatography is a specialized sample introduction technique in which a high molecular weight sample is heated rapidly to pyrolysing temperatures up to 900°C in a nonoxidizing atmosphere
  • SI- (N&P) Needle Kit
Specifications
Description SI-1 (N&P) Needle Kit