Learn More
Sigma Aldrich Pyromellitic Dianhydride (pmda) - Reduced Graphene Oxide Composites For Lithium Ion Battery
Catalog No. 9213781G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
92-137-81G 1 g
1 options
Catalog No. 92-137-81G Supplier Sigma Aldrich Supplier No. 9213781G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Polyimide (PI) is a novel organic material for new generation energy storage devices. PI can be a potential high-power electrode material for rechargeable battery and super-capacitor applications because its faster redox kinetics than inorganic materials. PI-graphene nanocomposite takes advantages of graphene's excellent electronic conductivity and high surface area, which addresses the issues of PI's electronic insulation and further improve its utilization. With the added benefit of high safety, flexibility and easy processability, PI composited graphene nanocomposites would penetrate the field of energy storage in the near future.
Specifications
Linear Formula CxHyOzNw
Quantity 1 g